tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR922539-1G OR922539-5G
APOSOR922539-1GEA 97.3 EUR
APOSOR922539-1G APOSOR922539-5G
tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate 97%
tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate
Formel: C₁₁H₂₁NO₃
Molecular Weight: 215,289 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16497040
CAS-Nummer: 208046-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR