4-Methoxypyrimidine-2-carbonitrile 95%

Lieferant: Apollo Scientific

OR922542-1G OR922542-250MG
APOSOR922542-1GEA 208 EUR
APOSOR922542-1G APOSOR922542-250MG
4-Methoxypyrimidine-2-carbonitrile 95%
4-Methoxypyrimidine-2-carbonitrile
Formel: C₆H₅N₃O
Molecular Weight: 135,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13705422
CAS-Nummer: 94789-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR