tert-Butyl 3-ethylideneazetidine-1-carboxylate
Lieferant: Apollo Scientific
OR922789-10G
OR922789-1G
OR922789-5G
APOSOR922789-10GEA
1150
EUR
APOSOR922789-10G
APOSOR922789-1G
APOSOR922789-5G
tert-Butyl 3-ethylideneazetidine-1-carboxylate
tert-Butyl 3-ethylideneazetidine-1-carboxylate
Formel:
C₁₀H₁₇NO₂ Molecular Weight: 183,247 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12031238 CAS-Nummer: 934665-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...