Methyl 2,5-dimethyl-3-furancarboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2,5-dimethylfuran-3-carboxylate
OR922882-100G
OR922882-25G
APOSOR922882-100GEA
1010
EUR
APOSOR922882-100G
APOSOR922882-25G
Methyl 2,5-dimethyl-3-furancarboxylate 95%
Methyl 2,5-dimethyl-3-furancarboxylate
Formel:
C₈H₁₀O₃ Molecular Weight: 154,163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010303 CAS-Nummer: 6148-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...