5-(tert-Butyl)-2-methylbenzoxazole 95%

Lieferant: Apollo Scientific

OR923126-1G OR923126-5G
APOSOR923126-1GEA 261 EUR
APOSOR923126-1G APOSOR923126-5G
5-(tert-Butyl)-2-methylbenzoxazole 95%
5-(tert-Butyl)-2-methylbenzoxazole
Formel: C₁₂H₁₅NO
Molecular Weight: 189,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027425
CAS-Nummer: 40874-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR