Methyl 6-methylpyrimidine-4-carboxylate 95%

Lieferant: Apollo Scientific

OR923171-1G OR923171-250MG
APOSOR923171-1GEA 109 EUR
APOSOR923171-1G APOSOR923171-250MG
Methyl 6-methylpyrimidine-4-carboxylate 95%
Methyl 6-methylpyrimidine-4-carboxylate
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13174781
CAS-Nummer: 73955-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR