N-Methyl-7h-pyrrolo[2,3-d]pyrimidin-4-amine 95%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine

OR923172-1G OR923172-5G
APOSOR923172-1GEA 33.5 EUR
APOSOR923172-1G APOSOR923172-5G
N-Methyl-7h-pyrrolo[2,3-d]pyrimidin-4-amine 95%
N-Methyl-7h-pyrrolo[2,3-d]pyrimidin-4-amine
Formel: C₇H₈N₄
Molecular Weight: 148,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01346677
CAS-Nummer: 78727-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR