4-[(2-Methyl-2-propen-1-yl)oxy]benzaldehyde 95%

Lieferant: Apollo Scientific

OR923263-1G OR923263-5G
APOSOR923263-1GEA 168 EUR
APOSOR923263-1G APOSOR923263-5G
4-[(2-Methyl-2-propen-1-yl)oxy]benzaldehyde 95%
4-[(2-Methyl-2-propen-1-yl)oxy]benzaldehyde
Formel: C₁₁H₁₂O₂
Molecular Weight: 176.212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00845682
CAS-Nummer: 38002-91-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR