Pyridin-4-yl-acetic acid tert-butyl ester 95%

Lieferant: Apollo Scientific

OR923273-100MG OR923273-1G OR923273-250MG
APOSOR923273-100MGEA 161 EUR
APOSOR923273-100MG APOSOR923273-1G APOSOR923273-250MG
Pyridin-4-yl-acetic acid tert-butyl ester 95%
Pyridin-4-yl-acetic acid tert-butyl ester
Formel: C₁₁H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22565620
CAS-Nummer: 79757-20-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR