2-Oxa-8-azaspiro[4.5]decane 95%

Lieferant: Apollo Scientific

OR923358-1G OR923358-250MG OR923358-100MG
APOSOR923358-1GEA 633 EUR
APOSOR923358-1G APOSOR923358-250MG APOSOR923358-100MG
2-Oxa-8-azaspiro[4.5]decane 95%
2-Oxa-8-azaspiro[4.5]decane
Formel: C₈H₁₅NO
Molecular Weight: 141,211 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11035890
CAS-Nummer: 176-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR