5,6,7,8-Tetrahydro-3H-quinazolin-4-one 95%

Lieferant: Apollo Scientific

OR923476-1G OR923476-5G
APOSOR923476-1GEA 190 EUR
APOSOR923476-1G APOSOR923476-5G
5,6,7,8-Tetrahydro-3H-quinazolin-4-one 95%
5,6,7,8-Tetrahydro-3H-quinazolin-4-one
Formel: C₈H₁₀N₂O
Molecular Weight: 150,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01749933
CAS-Nummer: 19178-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR