3-(1H-1,2,4-Triazol-1-ylmethyl)benzaldehyde 97%

Lieferant: Apollo Scientific
Gefahr

OR9236-1G OR9236-250MG OR9236-5G
APOSOR9236-1GEA 248 EUR
APOSOR9236-1G APOSOR9236-250MG APOSOR9236-5G
3-(1H-1,2,4-Triazol-1-ylmethyl)benzaldehyde 97%
3-(1H-1,2,4-Triazol-1-ylmethyl)benzaldehyde
Formel: C₁₀H₉N₃O
Molecular Weight: 187,198 g/mol
Schmelzpunkt: 54…55,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08271928
CAS-Nummer: 876316-30-2
UN: 1325
ADR: 4.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR