2-(Benzofuran-5-yl)acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-(Benzofuran-5-yl)acetic Acid

OR923851-1G OR923851-250MG
APOSOR923851-1GEA 358 EUR
APOSOR923851-1G APOSOR923851-250MG
2-(Benzofuran-5-yl)acetic acid 95%
2-(Benzofuran-5-yl)acetic acid
Formel: C₁₀H₈O₃
Molecular Weight: 176,169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09925119
CAS-Nummer: 142935-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR