(2E)-3-(4-Chlorophenyl)acrylonitrile 95%

Lieferant: Apollo Scientific

OR923905-10G OR923905-1G OR923905-5G
APOSOR923905-10GEA 400 EUR
APOSOR923905-10G APOSOR923905-1G APOSOR923905-5G
(2E)-3-(4-Chlorophenyl)acrylonitrile 95%
(2E)-3-(4-Chlorophenyl)acrylonitrile
Formel: C₉H₆ClN
Molecular Weight: 163,604 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001932
CAS-Nummer: 28446-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR