2-Ethynyl-6-methylpyridine 95%

Lieferant: Apollo Scientific

OR924008-1G OR924008-250MG
APOSOR924008-1GEA 282 EUR
APOSOR924008-1G APOSOR924008-250MG
2-Ethynyl-6-methylpyridine 95%
2-Ethynyl-6-methylpyridine
Formel: C₈H₇N
Molecular Weight: 117.148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08703491
CAS-Nummer: 30413-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR