trans-3-Hydroxycyclobutanecarboxylic acid

Lieferant: Apollo Scientific

OR924057-1G OR924057-5G
APOSOR924057-1GEA 293 EUR
APOSOR924057-1G APOSOR924057-5G
trans-3-Hydroxycyclobutanecarboxylic acid
trans-3-Hydroxycyclobutanecarboxylic acid
Formel: C₅H₈O₃
Molecular Weight: 116,115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17676395
CAS-Nummer: 1268521-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR