1-(5-Amino-1H-indol-1-yl)ethanone 95%
Lieferant: Apollo Scientific
Synonyme:
1-Acetyl-5-aminoindole
OR924110-1G
OR924110-250MG
APOSOR924110-1GEA
230
EUR
APOSOR924110-1G
APOSOR924110-250MG
1-(5-Amino-1H-indol-1-yl)ethanone 95%
1-(5-Amino-1H-indol-1-yl)ethanone
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174,199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11559037 CAS-Nummer: 16066-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...