2-(3-Methoxypyridin-2-yl)acetic acid 96%

Lieferant: Apollo Scientific

OR924160-1G OR924160-250MG
APOSOR924160-1GEA 632 EUR
APOSOR924160-1G APOSOR924160-250MG
2-(3-Methoxypyridin-2-yl)acetic acid 96%
2-(3-Methoxypyridin-2-yl)acetic acid
Formel: C₈H₉NO₃
Molecular Weight: 167.162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09924953
CAS-Nummer: 1000515-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR