5-(Methylsulphonyl)-1H-indazole 95%
Lieferant: Apollo Scientific
OR924191-1G
OR924191-250MG
APOSOR924191-1GEA
754
EUR
APOSOR924191-1G
APOSOR924191-250MG
5-(Methylsulphonyl)-1H-indazole 95%
5-(Methylsulphonyl)-1H-indazole
Formel:
C₈H₈N₂O₂S Molecular Weight: 196.226 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13190321 CAS-Nummer: 1268816-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...