Ethyl indane-5-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl Indane-5-carboxylate

OR924196-1G OR924196-250MG
APOSOR924196-1GEA 368 EUR
APOSOR924196-1G APOSOR924196-250MG
Ethyl indane-5-carboxylate 95%
Ethyl indane-5-carboxylate
Formel: C₁₂H₁₄O₂
Molecular Weight: 190,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12179700
CAS-Nummer: 105640-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR