Dimethyl 3-aminophthalate 95%

Lieferant: Apollo Scientific
Achtung

OR924260-100G OR924260-25G OR924260-5G
APOSOR924260-100GEA 642 EUR
APOSOR924260-100G APOSOR924260-25G APOSOR924260-5G
Dimethyl 3-aminophthalate 95%
Dimethyl 3-aminophthalate
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209,199 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09029586
CAS-Nummer: 34529-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR