5-Ethylpyrimidin-2-amine 95%

Lieferant: Apollo Scientific

OR924263-1G OR924263-250MG
APOSOR924263-1GEA 302 EUR
APOSOR924263-1G APOSOR924263-250MG
5-Ethylpyrimidin-2-amine 95%
5-Ethylpyrimidin-2-amine
Formel: C₆H₉N₃
Molecular Weight: 123,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09743848
CAS-Nummer: 39268-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR