Ethyl 1-methyl-3-oxocyclobutane-1-carboxylate
Lieferant: Apollo Scientific
OR924414-1G
OR924414-25G
OR924414-5G
APOSOR924414-1GEA
265
EUR
APOSOR924414-1G
APOSOR924414-25G
APOSOR924414-5G
Ethyl 1-methyl-3-oxocyclobutane-1-carboxylate
Ethyl 1-methyl-3-oxocyclobutane-1-carboxylate
Formel:
C₈H₁₂O₃ Molecular Weight: 156,179 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23105920 CAS-Nummer: 227607-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...