N,N-Diethylthiophene-3-carboxamide 96%

Lieferant: Apollo Scientific

OR924466-10G OR924466-1G OR924466-25G
APOSOR924466-10GEA 128 EUR
APOSOR924466-10G APOSOR924466-1G APOSOR924466-25G
N,N-Diethylthiophene-3-carboxamide 96%
N,N-Diethylthiophene-3-carboxamide
Formel: C₉H₁₃NOS
Molecular Weight: 183.271 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11742083
CAS-Nummer: 73540-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR