3-Amino-2-bromophenol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-amino-2-bromophenol
OR924467-1G
OR924467-250MG
APOSOR924467-1GEA
53.8
EUR
APOSOR924467-1G
APOSOR924467-250MG
3-Amino-2-bromophenol 95+%
3-Amino-2-bromophenol
Formel:
C₆H₆BrNO Molecular Weight: 188,022 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846453 CAS-Nummer: 100367-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...