(1R,3R,5R)-2-[(2-Methylpropan-2-yl)oxycarbonyl]-2-azabicyclo[3.1.0]hexane-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1R,3R,5R)-2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-3-carboxylic acid

OR924521-250MG OR924521-500MG
APOSOR924521-250MGEA 231 EUR
APOSOR924521-250MG APOSOR924521-500MG
(1R,3R,5R)-2-[(2-Methylpropan-2-yl)oxycarbonyl]-2-azabicyclo[3.1.0]hexane-3-carboxylic acid 97%
(1R,3R,5R)-2-[(2-Methylpropan-2-yl)oxycarbonyl]-2-azabicyclo[3.1.0]hexane-3-carboxylic acid
Formel: C₁₁H₁₇NO₄
Lagertemperatur: Kühlschrank
MDL: MFCD16621713
CAS-Nummer: 1148048-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR