Methyl 5-methylpyrimidine-2-carboxylate 95%
Lieferant: Apollo Scientific
OR924529-1G
OR924529-250MG
APOSOR924529-1GEA
177
EUR
APOSOR924529-1G
APOSOR924529-250MG
Methyl 5-methylpyrimidine-2-carboxylate 95%
Methyl 5-methylpyrimidine-2-carboxylate
Formel:
C₇H₈N₂O₂ Molecular Weight: 152,151 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16660966 CAS-Nummer: 76196-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...