1-Boc-2,5-dihydro-3-methyl-1H-pyrrole 95%

Lieferant: Apollo Scientific

OR924534-100MG OR924534-1G OR924534-250MG
APOSOR924534-100MGEA 183 EUR
APOSOR924534-100MG APOSOR924534-1G APOSOR924534-250MG
1-Boc-2,5-dihydro-3-methyl-1H-pyrrole 95%
1-Boc-2,5-dihydro-3-methyl-1H-pyrrole
Formel: C₁₀H₁₇NO₂
Molecular Weight: 183,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24466973
CAS-Nummer: 643759-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR