Pyridazine-4-carbonitrile

Lieferant: Apollo Scientific

OR924578-1G OR924578-5G
APOSOR924578-1GEA 476 EUR
APOSOR924578-1G APOSOR924578-5G
Pyridazine-4-carbonitrile
Pyridazine-4-carbonitrile
Formel: C₅H₃N₃
Molecular Weight: 105,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263328
CAS-Nummer: 68776-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR