2-Amino-3-(biphenyl-4-yl)propanoic acid 97%
Lieferant: Apollo Scientific
Synonyme: 2-Amino-3-(4-biphenylyl)propanoic Acid
OR924588-1G OR924588-5G
APOSOR924588-1GEA 127 EUR
APOSOR924588-1G APOSOR924588-5G
2-Amino-3-(biphenyl-4-yl)propanoic acid 97%
2-Amino-3-(biphenyl-4-yl)propanoic acid
Formel: C₁₅H₁₅NO₂
Molecular Weight: 241.285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425234
CAS-Nummer: 76985-08-5