Ethyl 2-(1H-imidazol-1-yl)-2-oxoacetate 95%
Lieferant: Apollo Scientific
OR924600-1G
OR924600-5G
APOSOR924600-1GEA
219
EUR
APOSOR924600-1G
APOSOR924600-5G
Ethyl 2-(1H-imidazol-1-yl)-2-oxoacetate 95%
Ethyl 2-(1H-imidazol-1-yl)-2-oxoacetate
Formel:
C₇H₈N₂O₃ Molecular Weight: 168,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21362621 CAS-Nummer: 75716-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...