Methyl 3-hydroxy-1-methylcyclobutane-1-carboxylate

Lieferant: Apollo Scientific

OR924751-10G OR924751-1G OR924751-5G
APOSOR924751-10GEA 2050 EUR
APOSOR924751-10G APOSOR924751-1G APOSOR924751-5G
Methyl 3-hydroxy-1-methylcyclobutane-1-carboxylate
Methyl 3-hydroxy-1-methylcyclobutane-1-carboxylate
Formel: C₇H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD23105985
CAS-Nummer: 169899-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR