2-(Methoxymethyl)azetidine

Lieferant: Apollo Scientific

OR924801-10G OR924801-1G OR924801-5G
APOSOR924801-10GEA 2020 EUR
APOSOR924801-10G APOSOR924801-1G APOSOR924801-5G
2-(Methoxymethyl)azetidine
2-(Methoxymethyl)azetidine
Formel: C₅H₁₁NO
Molecular Weight: 101.147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105901
CAS-Nummer: 1290136-94-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR