1-Amino-4-bromo-1H-pyrrole-2-carbonitrile 95%

Lieferant: Apollo Scientific

OR924899-1G OR924899-250MG
APOSOR924899-1GEA 828 EUR
APOSOR924899-1G APOSOR924899-250MG
1-Amino-4-bromo-1H-pyrrole-2-carbonitrile 95%
1-Amino-4-bromo-1H-pyrrole-2-carbonitrile
Formel: C₅H₄BrN₃
Molecular Weight: 186.009 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105808
CAS-Nummer: 937161-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR