4,6-Dimethoxypyrimidine-5-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR924951-1G OR924951-25G OR924951-5G
APOSOR924951-1GEA 29 EUR
APOSOR924951-1G APOSOR924951-25G APOSOR924951-5G
4,6-Dimethoxypyrimidine-5-carboxylic acid 97%
4,6-Dimethoxypyrimidine-5-carboxylic acid
Formel: C₇H₈N₂O₄
Molecular Weight: 184.149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11557227
CAS-Nummer: 4319-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR