Methyl isoquinoline-5-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-Isoquinolinecarboxylate

OR925357-1G
APOSOR925357-1GEA 60.9 EUR
APOSOR925357-1G
Methyl isoquinoline-5-carboxylate 95%
Methyl isoquinoline-5-carboxylate
Formel: C₁₁H₉NO₂
Molecular Weight: 187,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06204278
CAS-Nummer: 16675-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR