3'-Hydroxypropiophenone 97%
Lieferant: Apollo Scientific
Synonyme:
1-(3-Hydroxyphenyl)propan-1-one
OR925473-100G
APOSOR925473-100GEA
132
EUR
APOSOR925473-100G
3'-Hydroxypropiophenone 97%
3'-Hydroxypropiophenone
Formel:
C₉H₁₀O₂ Molecular Weight: 150,175 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03428514 CAS-Nummer: 13103-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...