2-[(Boc-amino)methyl]benzoic acid 97%

Lieferant: Apollo Scientific

OR925728-1G OR925728-500MG
APOSOR925728-1GEA 393 EUR
APOSOR925728-1G APOSOR925728-500MG
2-[(Boc-amino)methyl]benzoic acid 97%
2-[(Boc-amino)methyl]benzoic acid
Formel: C₁₃H₁₇NO₄
Molecular Weight: 251,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10565780
CAS-Nummer: 669713-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR