3-(Aminomethyl)-1-benzylpyrazole dihydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: (1-Benzylpyrazol-3-yl)methanamine dihydrochloride

OR925794-100MG OR925794-250MG
APOSOR925794-100MGEA 143 EUR
APOSOR925794-100MG APOSOR925794-250MG
3-(Aminomethyl)-1-benzylpyrazole dihydrochloride 95%
3-(Aminomethyl)-1-benzylpyrazole dihydrochloride
Formel: C₁₁H₁₃N₃·2ClH
Molecular Weight: 260,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23703523
CAS-Nummer: 1427475-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR