4-Hydroxy-7-methoxy-1H-quinolin-2-one 95%

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-7-methoxyquinolin-2(1H)-one

OR925894-1G OR925894-5G
APOSOR925894-1GEA 78.3 EUR
APOSOR925894-1G APOSOR925894-5G
4-Hydroxy-7-methoxy-1H-quinolin-2-one 95%
4-Hydroxy-7-methoxy-1H-quinolin-2-one
Formel: C₁₀H₉NO₃
Molecular Weight: 191,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00101261
CAS-Nummer: 27037-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR