(5-Chloro-2-thienyl)acetic acid 95%

Lieferant: Apollo Scientific

OR925923-100MG OR925923-1G OR925923-250MG
APOSOR925923-100MGEA 168 EUR
APOSOR925923-100MG APOSOR925923-1G APOSOR925923-250MG
(5-Chloro-2-thienyl)acetic acid 95%
(5-Chloro-2-thienyl)acetic acid
Formel: C₆H₅ClO₂S
Molecular Weight: 176,621 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01859875
CAS-Nummer: 13669-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR