4-Ethyl-3,5-dimethyl-1H-pyrrole-2-carbaldehyde 95%
Lieferant: Apollo Scientific
OR925987-1G
OR925987-5G
APOSOR925987-1GEA
139
EUR
APOSOR925987-1G
APOSOR925987-5G
4-Ethyl-3,5-dimethyl-1H-pyrrole-2-carbaldehyde 95%
4-Ethyl-3,5-dimethyl-1H-pyrrole-2-carbaldehyde
Formel:
C₉H₁₃NO Molecular Weight: 151,206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864532 CAS-Nummer: 6250-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...