N-Ethyl-2-piperidin-1-ylethanamine 95%
Lieferant: Apollo Scientific
OR926190-1G
OR926190-250MG
APOSOR926190-1GEA
364
EUR
APOSOR926190-1G
APOSOR926190-250MG
N-Ethyl-2-piperidin-1-ylethanamine 95%
N-Ethyl-2-piperidin-1-ylethanamine
Formel:
C₉H₂₀N₂ Molecular Weight: 156.269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08361609 CAS-Nummer: 32776-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...