6-Methoxyquinazolin-4-ol 98%
Lieferant: Apollo Scientific
OR926223-1G
OR926223-5G
APOSOR926223-1GEA
62.5
EUR
APOSOR926223-1G
APOSOR926223-5G
6-Methoxyquinazolin-4-ol 98%
6-Methoxyquinazolin-4-ol
Formel:
C₉H₈N₂O₂ Molecular Weight: 176,172 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657614 CAS-Nummer: 19181-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...