4-(Chlorosulphonyl)phenyl pivalate 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(4-Chlorosulphonylphenyl) 2,2-dimethylpropanoate
OR926337-1G
OR926337-250MG
APOSOR926337-1GEA
200
EUR
APOSOR926337-1G
APOSOR926337-250MG
4-(Chlorosulphonyl)phenyl pivalate 95%
4-(Chlorosulphonyl)phenyl pivalate
Formel:
C₁₁H₁₃ClO₄S Molecular Weight: 276.737 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08274725 CAS-Nummer: 150374-99-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...