2-(3-Methyl-2-nitrophenyl)acetonitrile 95%

Lieferant: Apollo Scientific

OR926415-1G OR926415-250MG
APOSOR926415-1GEA 435 EUR
APOSOR926415-1G APOSOR926415-250MG
2-(3-Methyl-2-nitrophenyl)acetonitrile 95%
2-(3-Methyl-2-nitrophenyl)acetonitrile
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08276005
CAS-Nummer: 91192-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR