6-Chloro-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid, N-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-chloro-2-[(2-methylpropan-2-yl)oxycarbonyl]-3,4-dihydro-1H-isoquinoline-1-carboxylic acid

OR926499-250MG OR926499-1G
APOSOR926499-250MGEA 437 EUR
APOSOR926499-250MG APOSOR926499-1G
6-Chloro-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid, N-BOC protected
6-Chloro-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid, N-BOC protected
Formel: C₁₅H₁₈ClNO₄
Molecular Weight: 311.761 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114850
CAS-Nummer: 871730-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR