Ethyl 4,5,6,7-tetrahydro-1H-indazole-5-carboxylate 95%

Lieferant: Apollo Scientific

OR926624-1G OR926624-250MG
APOSOR926624-1GEA 108 EUR
APOSOR926624-1G APOSOR926624-250MG
Ethyl 4,5,6,7-tetrahydro-1H-indazole-5-carboxylate 95%
Ethyl 4,5,6,7-tetrahydro-1H-indazole-5-carboxylate
Formel: C₁₀H₁₄N₂O₂
Molecular Weight: 194,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21364708
CAS-Nummer: 792848-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR