3-Methyl-5-propyl-cyclohex-2-en-1-one 95%
Lieferant: Apollo Scientific
OR926643-1G
OR926643-5G
APOSOR926643-1GEA
226
EUR
APOSOR926643-1G
APOSOR926643-5G
3-Methyl-5-propyl-cyclohex-2-en-1-one 95%
3-Methyl-5-propyl-cyclohex-2-en-1-one
Formel:
C₁₀H₁₆O Molecular Weight: 152,233 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00482082 CAS-Nummer: 3720-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...