(S)-3-Piperidinecarboxylic acid methyl ester hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme: (S)-Methyl piperidine-3-carboxylate hydrochloride
OR926692-1G OR926692-5G
APOSOR926692-1GEA 60.9 EUR
APOSOR926692-1G APOSOR926692-5G
(S)-3-Piperidinecarboxylic acid methyl ester hydrochloride 95%
(S)-3-Piperidinecarboxylic acid methyl ester hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09271813
CAS-Nummer: 164323-84-6